AC5H30533AAA Accela ChemBio Inc.

Cyanocobalamin

  • MDL number MFCD00151092
  • Cas Number : 68-19-9
  • Molecular Weight : 1355.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010769&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY010769.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart