AC5H30533ACC Accela ChemBio Inc.

Disodium 4-Formylbenzene-1,3-disulfonate

  • MDL number MFCD00043913
  • Cas Number : 33513-44-9
  • Molecular Weight : 310.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016484&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016484.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart