AC5H30533AE4 Accela ChemBio Inc.

1-[Bis(dimethylamino)methylene]-5-chloro-1-benzotriazolium 3-Oxide Tetrafluoroborate

  • MDL number MFCD08461912
  • Cas Number : 330641-16-2
  • Molecular Weight : 355.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019130&attname=GoodsCode

Safety Information

signalword Danger
pcodes P260-P264-P270-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363
hcodes GHS05,GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY019130.pdf
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart