AC5H30533B25 Accela ChemBio Inc.

2-(4-Amino-2-pyridyl)ethanol

  • MDL number MFCD18816527
  • Cas Number : 101012-38-8
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026069&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026069.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart