AC5H30533B36 Accela ChemBio Inc.

4-Methoxypyridine-3-boronic Acid Pinacol Ester

  • MDL number MFCD07367561
  • Cas Number : 758699-74-0
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028915&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028915.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart