AC5H30533B37 Accela ChemBio Inc.

trans-2-Phenylcyclopropanecarboxylic Acid

  • MDL number MFCD00001292
  • Cas Number : 939-90-2
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028926&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028926.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart