AC5H30533B71 Accela ChemBio Inc.

1-Boc-4-[5-(aminomethyl)-2-pyrimidyl]piperazine

  • MDL number MFCD10696917
  • Cas Number : 944899-79-0
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035242&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035242.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart