AC5H30533B77 Accela ChemBio Inc.

(Fluoromethyl)triphenylphosphonium Tetrafluoroborate

  • MDL number MFCD22124052
  • Cas Number : 96385-23-8
  • Molecular Weight : 382.115057

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036077&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY036077.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart