AC5H30533B8E Accela ChemBio Inc.

Isophthalohydrazide

  • MDL number MFCD00025117
  • Cas Number : 2760-98-7
  • Molecular Weight : 194.1906

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040656&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY040656.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart