AC5H30533B8F Accela ChemBio Inc.

5-Methylisoxazole-4-carboxamide

  • MDL number MFCD11643943
  • Cas Number : 1097817-28-1
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040666&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040666.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart