AC5H30533B91 Accela ChemBio Inc.

6-Amino-5-fluoropyridine-3-boronic Acid Pinacol Ester

  • MDL number MFCD12923417
  • Cas Number : 944401-75-6
  • Molecular Weight : 238.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040961&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040961.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart