AC5H30533BBF Accela ChemBio Inc.

(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran

  • MDL number MFCD27920793
  • Cas Number : 915095-89-5
  • Molecular Weight : 367.66474

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY046908&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY046908.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart