AC5H30533BD3 Accela ChemBio Inc.

Methyl (Methylsulfinyl)methyl Sulfide

  • MDL number MFCD00002091
  • Cas Number : 33577-16-1
  • Molecular Weight : 124.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY049552&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY049552.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart