AC5H30533BE8 Accela ChemBio Inc.

7-(Bromomethyl)pentadecane

  • MDL number MFCD00136231
  • Cas Number : 52997-43-0
  • Molecular Weight : 305.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY055275&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY055275.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart