AC5H30533BEC Accela ChemBio Inc.

2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine

  • MDL number MFCD00194632
  • Cas Number : 23449-08-3
  • Molecular Weight : 388.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY055731&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY055731.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart