AC5H30533BED Accela ChemBio Inc.

4-Nitrotriphenylamine

  • MDL number MFCD00814232
  • Cas Number : 4316-57-8
  • Molecular Weight : 290.31596

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY056308&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY056308.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart