AC5H30533BF0 Accela ChemBio Inc.

Thieno[2,3-b]pyridine 1,1-Dioxide

  • MDL number MFCD18803426
  • Cas Number : 37049-39-1
  • Molecular Weight : 167.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY056751&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY056751.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart