AC5H30533C37 Accela ChemBio Inc.

5-Nitro BAPTA Tetramethyl Ester

  • MDL number MFCD28358710
  • Cas Number : 172646-43-4
  • Molecular Weight : 577.53724

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY073580&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY073580.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart