AC5H30533C45 Accela ChemBio Inc.

2,3,6,7-Tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one

  • MDL number MFCD09743876
  • Cas Number : 61196-37-0
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY083738&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY083738.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart