AC5H30533C47 Accela ChemBio Inc.

N-(2-Hydroxy-4-nitrophenyl)acetamide

  • MDL number MFCD00219839
  • Cas Number : 25351-89-7
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY093784&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY093784.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart