AC5H30533C52 Accela ChemBio Inc.

1,2,3,4-Tetrahydroisoquinoline-6-carboxylic Acid

  • MDL number MFCD09027511
  • Cas Number : 933752-32-0
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY100958&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY100958.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart