AC5H30533C5C Accela ChemBio Inc.

3-(2-Methoxy-2-oxoethyl)phenylboronic Acid Pinacol Ester

  • MDL number MFCD13195752
  • Cas Number : 478375-42-7
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY102390&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY102390.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart