AC5H30533C78 Accela ChemBio Inc.

Isobutyltriphenylphosphonium Bromide

  • MDL number MFCD00031593
  • Cas Number : 22884-29-3
  • Molecular Weight : 399.303721

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY106402&attname=GoodsCode

Safety Information

pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
hcodes GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY106402.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart