AC5H30533C96 Accela ChemBio Inc.

1-(2-Hydroxyethyl)-1H-tetrazole-5(2H)-thione

  • MDL number MFCD06658157
  • Cas Number : 56610-81-2
  • Molecular Weight : 146.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY109999&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY109999.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart