AC5H30533CA2 Accela ChemBio Inc.

3-Aminodihydrofuran-2(3H)-one Hydrochloride

  • MDL number MFCD27977179
  • Cas Number : 42417-39-0
  • Molecular Weight : 137.56482

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY111957&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY111957.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart