AC5H30533CA9 Accela ChemBio Inc.

6-Bromo-1H-indole-3-ethanamine Hydrochloride

  • MDL number MFCD11109987
  • Cas Number : 108061-77-4
  • Molecular Weight : 275.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY112831&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY112831.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart