AC5H30533CB2 Accela ChemBio Inc.

(R)-4-Amino-3-(4-phenoxyphenyl)-1-(3-piperidyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD20482137
  • Cas Number : 1022150-12-4
  • Molecular Weight : 386.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY114940&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY114940.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart