AC5H30533CBE Accela ChemBio Inc.

L-Threonine Benzyl Ester Hydrochloride

  • MDL number MFCD00038977
  • Cas Number : 33645-24-8
  • Molecular Weight : 245.70264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY120978&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY120978.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart