AC5H30533CC7 Accela ChemBio Inc.

4-(Trifluoromethyl)tetrahydropyran-4-ol

  • MDL number MFCD16067947
  • Cas Number : 1251358-35-6
  • Molecular Weight : 170.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY123532&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY123532.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart