AC5H30533CCA Accela ChemBio Inc.

3-(Boc-amino)-1-[(tert-butyldimethylsilyl)oxy]-2-propanone

  • MDL number MFCD24466259
  • Cas Number : 184429-84-3
  • Molecular Weight : 303.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY124739&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY124739.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart