AC5H30533CCC Accela ChemBio Inc.

4-(1H-1,2,3-Triazol-1-yl)phenylboronic Acid Pinacol Ester

  • MDL number MFCD22570712
  • Cas Number : 1101174-00-8
  • Molecular Weight : 271.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY125640&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY125640.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart