AC5H30533CD0 Accela ChemBio Inc.

5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine

  • MDL number MFCD13705330
  • Cas Number : 65541-95-9
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY128852&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY128852.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart