AC5H30533CD1 Accela ChemBio Inc.

6-Aminobenzoisothiazole

  • MDL number MFCD18804902
  • Cas Number : 97050-36-7
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129143&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129143.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart