AC5H30533CD4 Accela ChemBio Inc.

1,6-Naphthyridin-2(1H)-one

  • MDL number MFCD07781179
  • Cas Number : 23616-29-7
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129309&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129309.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart