AC5H30533CD5 Accela ChemBio Inc.

6-Bromo-5-fluoroisatin

  • MDL number MFCD28678391
  • Cas Number : 1374208-41-9
  • Molecular Weight : 244.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129332&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129332.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart