AC5H30533CD6 Accela ChemBio Inc.

6-Aminobenzo[c]isothiazole

  • MDL number MFCD21145734
  • Cas Number : 1440512-64-0
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129380&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129380.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart