AC5H30533CD9 Accela ChemBio Inc.

[1,2,4]Triazolo[1,5-a]pyridine-5-boronic Acid Pinacol Ester

  • MDL number MFCD13181996
  • Cas Number : 2009345-74-6
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129664&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129664.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart