AC5H30533CDB Accela ChemBio Inc.

Oxazole-5-acetic Acid

  • MDL number MFCD22071442
  • Cas Number : 1083337-90-9
  • Molecular Weight : 127.0981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129690&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129690.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart