AC5H30533CDE Accela ChemBio Inc.

7-Fluorobenzoxazol-2(3H)-one

  • MDL number MFCD11845133
  • Cas Number : 509147-83-5
  • Molecular Weight : 153.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY130165&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY130165.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart