AC5H30533CE2 Accela ChemBio Inc.

Methyl (2,2,2-Trifluoroethyl) Carbonate

  • MDL number MFCD28054295
  • Cas Number : 156783-95-8
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY131079&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY131079.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart