AC5H30533CE3 Accela ChemBio Inc.

Methyl (S)-2-Amino-3-(3-bromophenyl)propanoate

  • MDL number MFCD08058222
  • Cas Number : 875782-96-0
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY131080&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY131080.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart