AC5H30533CF3 Accela ChemBio Inc.

2-(Phenylsulfonyl)ethanamine Hydrochloride

  • MDL number MFCD03840163
  • Cas Number : 38752-48-6
  • Molecular Weight : 221.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY173698&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY173698.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart