AC5H30533CFC Accela ChemBio Inc.

2-Aminothieno[2,3-d]thiazole

  • MDL number MFCD18451327
  • Cas Number : 40507-56-0
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY211665&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY211665.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart