AC5H30533CFD Accela ChemBio Inc.

Methyl 3-Cyclopropylpropiolate

  • MDL number MFCD03035227
  • Cas Number : 80866-48-4
  • Molecular Weight : 124.13722

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY214173&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY214173.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart