AC5H30533D06 Accela ChemBio Inc.

(S)-4-Phenyloxazolidine-2,5-dione

  • MDL number MFCD11978065
  • Cas Number : 3412-48-4
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY225275&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY225275.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart