AC5H30533D0E Accela ChemBio Inc.

5-Nitrobenzo[b]thiophen-3(2H)-one 1,1-Dioxide

  • MDL number MFCD01924626
  • Cas Number : 343348-19-6
  • Molecular Weight : 227.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226112&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226112.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart