AC5H30533D0F Accela ChemBio Inc.

Thieno[2,3-b]pyridin-3(2H)-one 1,1-Dioxide

  • MDL number MFCD31699884
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226113&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226113.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart