AC5H30533D10 Accela ChemBio Inc.

Ethyl 1H-Pyrazolo[3,4-d]pyrimidine-6-carboxylate

  • MDL number MFCD31630348
  • Cas Number : 2106690-78-0
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226290&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226290.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart