AC5H30533D11 Accela ChemBio Inc.

1,3:2,4-Dibenzylidene Sorbitol

  • MDL number MFCD01753099
  • Cas Number : 32647-67-9
  • Molecular Weight : 358.38508

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226316&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226316.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart