AC5H30533D15 Accela ChemBio Inc.

2-Nitro-8,9-dihydro-5H-benzo[7]annulen-7(6H)-one

  • MDL number MFCD11869793
  • Cas Number : 740842-50-6
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226858&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226858.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart